missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1-Fc Recombinant Human Protein, Invitrogen™ Sino Biological™
Click to view available options
Quantity:
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
250µg
25µg
50µg
Description
Immunoglobulin gamma-1 heavy chain constant region, partial (IgG1-Fc), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.&line-break;Background: Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.
- N-terminal Sequence Analysis: Glu 99
- Activity: 1. Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H1) in a functional ELISA; 2. Measured by its ability to bind human CD16a-AVI-His (Mfr. No. 10389-H27H) in a functional ELISA; 3.Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H) in a functional ELISA
- Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween™-80
- Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Molecular Weight (g/mol) | 26kDa |
| Quantity | 5 x 50 μg |
| Storage Requirements | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
| Structural Form | Truncated/Recombinant |
| Endotoxin Concentration | <1 EU/μg |
| Gene Alias | Fc region of human immunoglobulin G1 |
| Common Name | Fc region of human immunoglobulin G1 |
| Gene Symbol | IgG1-Fc |
| Protein Tag | IgG1 Fc Tag |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction