missing translation for 'onlineSavingsMsg'
Learn More

IgG1-Fc Recombinant Human Protein, Invitrogen™ Sino Biological™

Product Code. 13425946
Change view
Click to view available options
Quantity:
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
250µg
25µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13425946 5 x 5 μg 25µg
13455946 50 μg 50µg
13435946 5 x 50 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13425946 Supplier Invitrogen Supplier No. 10702HNAH25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This recombinant human protein is expressed from a DNA sequence encoding the human IgG1-Fc region (AAC82527.1), containing residues 99 to 330, including a 103 Cys/Ser mutation

Immunoglobulin gamma-1 heavy chain constant region, partial (IgG1-Fc), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.&line-break;Background: Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.
  • N-terminal Sequence Analysis: Glu 99
  • Activity: 1. Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H1) in a functional ELISA; 2. Measured by its ability to bind human CD16a-AVI-His (Mfr. No. 10389-H27H) in a functional ELISA; 3.Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H) in a functional ELISA
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween™-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Molecular Weight (g/mol) 26kDa
Quantity 5 x 5 μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias Fc region of human immunoglobulin G1
Common Name Fc region of human immunoglobulin G1
Gene Symbol IgG1-Fc
Protein Tag IgG1 Fc Tag
Expression System Mammalian
Protein Family Growth Factors & Receptors
Form Lyophilized
Protein Subtype Other Proteins
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.