missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human IDO1 (P14902-1) (Ala 2-Gly 403) fused to a polyhistidine tag at the N-terminus
Brand: Invitrogen 11650H07E50
This item is not returnable.
View return policy
Description
Indoleamine 2,3-Dioxygenase 1 (IDO1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
- Reconstitution: No reconstitution needed
This protein is manufactured by Sino Biological Inc.
Specifications
3620 | |
PAGE | |
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
IDO, INDO | |
IDO1 |
46kDa | |
50μg | |
Full Length/Recombinant | |
Indoleamine 2,3-Dioxygenase 1 |
For Research Use Only. Not for use in diagnostic procedures.