missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Anhui/1/2005), His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Pro 337) of influenza A virus, subtype H5N1 (A/Anhui/1/2005), HA1 subunit (ABD28180.1) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 11048V08H250
This item is not returnable.
View return policy
Description
Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: This recombinant protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
37.5kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Hemagglutinin |
50μg | |
Truncated/Recombinant/Cell Surface Protein | |
HA1 | |
HA |
For Research Use Only. Not for use in diagnostic procedures.