missing translation for 'onlineSavingsMsg'
Learn More

FTH1 Recombinant Human Protein, Invitrogen™ Sino Biological™

Product Code. 15276255
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15276255

Brand: Invitrogen 13217HNAE50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This recombinant protein was expressed from a DNA sequence encoding the human FTH1 (P02794) (Met 1-Ser183)

Ferritin, Heavy Polypeptide 1 (FTH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Gene ID (Entrez) 2495
Molecular Weight (g/mol) 21.2kDa
Purification Method PAGE
Quantity 50 μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias OK/SW-cl.84, FHC, FTH, FTHL6, PIG15, PLIF
Common Name Ferritin, heavy polypeptide 1
Gene Symbol FTH1
Expression System E. coli
Protein Family Other Proteins
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.