missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF10 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)
£59.50 - £776.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Gene ID (Entrez) | 2255 |
Molecular Weight (g/mol) | 19.6kDa |
Purification Method | PAGE |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15216955
|
Invitrogen
10573HNAE250 |
5 x 50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15226955
|
Invitrogen
10573HNAE5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15236955
|
Invitrogen
10573HNAE50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.- Activity: This recombinant protein has not been tested
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
19.6kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
FGF10 | |
FGF10 | |
E. coli | |
Lyophilized |
2255 | |
PAGE | |
Full Length/Recombinant | |
Fibroblast Growth Factor 10 | |
Untagged | |
Growth Factors & Receptors | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.