Learn More
EFNB2 (EphrinB2) Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 229) of human EFNB2 (NP_004084.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 10881H03H5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Ephrin-B2 (EFNB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNB4-His at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-25ng/mL, immobilized human EFNB4 at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-125ng/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50.3kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Ephrin-B2 | |
His Tag/IgG1 Fc Tag | |
Kinases & Inhibitors | |
Ephrins |
1948 | |
5μg | |
Truncated/Recombinant/Cell Surface Protein/Ligand | |
EFNB2, RP11-272L14.1, EPLG5, HTKL, Htk-L, LERK5, mgC126226, mgC126227, mgC126228 | |
EFNB2 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.