missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD40 isoform 1 (P27512-1) (Met 1-Arg 193) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 50324M03H250
Includes: Five vials of lyophilized protein
Description
CD40 antigen, TNF receptor superfamily member 5 (CD40), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to bind recombinant human CD40L in a functional ELISA
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
47kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
RP23-428M13.1, AI326936, Bp50, GP39, HIGM1, IGM, IMD3, T-BAM, TRAP, Tnfrsf5, p50 | |
CD40 |
5 x 50μg | |
<1 EU/μg | |
CD40 Antigen |
For Research Use Only. Not for use in diagnostic procedures.