missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD274 (PDL1) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™
Description
CD274 molecule (CD274), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
- Activity: Measured by its binding ability in a functional ELISA; immobilized human PDCD1/PD1 at 10μg/mL (100μg/well) can bind recombinant human CD274/PDL1-Fc chimera with a linear range of 0.02-0.4μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Molecular Weight (g/mol) | 52kDa |
| Quantity | 5 x 5 μg |
| Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
| Endotoxin Concentration | <1 EU/μg |
| Gene Alias | B7-H, B7H1, mgC142294, mgC142296, PD-L1, PDCD1L1, PDCD1LG1, PDL1 |
| Common Name | CD274 molecule |
| Gene Symbol | CD274 (PDL1) |
| Protein Tag | IgG1 Fc Tag |
| Expression System | Mammalian |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?