missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD209 Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™
Description
Cluster of Differentiation 209 (CD209), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
- Activity: This recombinant protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Gene ID (Entrez) | 30835 |
| Molecular Weight (g/mol) | 65.8kDa |
| Quantity | 50 μg |
| Storage Requirements | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
| Structural Form | Truncated/Recombinant/Cell Surface Protein |
| Endotoxin Concentration | <1 EU/μg |
| Gene Alias | CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, mgC129965, CD209 |
| Common Name | CD209 molecule |
| Gene Symbol | CD209 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?