missing translation for 'onlineSavingsMsg'
Learn More

ANGPT2 (ANG2) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human ANGPT2 (NP_001138.1) (Met 1-Phe 496) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10691H08H50

Product Code. 15217065

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Angiopoietin 2 (ANGPT2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind recombinant human Tie2-His in a functional ELISA; 2. Measured by its ability to bind recombinant human Tie2-Fch in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
50μg
<1 EU/μg
Angiopoietin 2
His Tag
Growth Factors & Receptors
Other Growth Factors
56.3kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
AGPT2, ANG2, ANGPT2
ANGPT2
Mammalian
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.