Learn More
Spectrum Labs™ Single-Use Process Reservoirs
Click to view available options
For Use With (Application):
KR2i, KMPi systems
Includes:
Inlet and outlet 1/4 in. x 3/8 in. C-FlexBuffer and vent 1/8 in. x 1/4 in. C-Flex4 dip tubes
Volume (Metric):
2 L
Description
- Tubes are cut and positioned to ensure that all of solution is directed into the module, and to prevent foaming
- Reservoirs have been intentionally crafted for seamless integration into SpectrumLabs™ TFF pump systems
Specifications
Specifications
| Material | Copolyester |
| For Use With (Application) | KR2i, KMPi systems |
| Includes | Inlet and outlet 1/4 in. x 3/8 in. C-FlexBuffer and vent 1/8 in. x 1/4 in. C-Flex4 dip tubes |
| Type | Single-Use TFF Reservoir |
| Volume (Metric) | 2 L |