Learn More
Spectrum Labs™ Single-Use Process Reservoirs
Product Code. 15885091
Click to view available options
For Use With (Application):
KR2i, KMPi systems
Includes:
Inlet and outlet 1/4 in. x 3/8 in. C-FlexBuffer and vent 1/8 in. x 1/4 in. C-Flex4 dip tubes
Volume (Metric):
2 L
Product Code. 15885091
Brand: Spectrum Labs™ ACBT2D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spectrum Labs™ Single-Use Process Reservoirs are designed with a combination of configured ports and tubes to support tangential flow filtration (TFF) applications.
  • Tubes are cut and positioned to ensure that all of solution is directed into the module, and to prevent foaming
  • Reservoirs have been intentionally crafted for seamless integration into SpectrumLabs™ TFF pump systems
Specifications
Material Copolyester
For Use With (Application) KR2i, KMPi systems
Includes Inlet and outlet 1/4 in. x 3/8 in. C-FlexBuffer and vent 1/8 in. x 1/4 in. C-Flex4 dip tubes
Type Single-Use TFF Reservoir
Volume (Metric) 2 L