Learn More
SHOWA™ 7540 Single Use Nitrile Gloves, Powder Free
Product Code. 15762937
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
Product Code. 15762937
Brand: SHOWA™ 754006XS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Light, silicon free, highly penetration safe chemical protective glove! High tear resistance!
  • Silicon free.
  • Electrostatic dissipative in use (ESD).
  • Textured finish on finger-tips to enhance grip
  • Second skin feel, softer texture and easy donning.
  • Powder-free and approved for food-handling.
  • Latex-free to avoid type I and type IV latex allergies.
  • Category III, EN 374 and EN 455 approved.
  • Suitable for food, pharmaceutical, aerospace, electronics and printing industries.
Specifications
Certifications/Compliance (Europe) EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011
AQL 0.65
Color Cobalt Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
EN ISO 374-1 Type C
EN ISO 374-1 CHEMICAL LETTER CODES JK
Glove Cuff Style Straight
Model SHO75400
Packaging Type dispenser box
Quantity 100/Pk.
Length (English) 9.5 in.
Length (Metric) 24 cm