Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Product Code. 16689561
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16689561 250 mL Each
16679561 125 mL Each
16699561 500 mL Each
16609571 1 L Each
16619571 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16689561 Supplier Masterflex™ Supplier No. 9564422
Add to basket
Add to basket
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Description Gamma Irradiated
Capacity (Metric) 250 mL