Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Each
Description
- Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
- Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Specifications
| Description | Gamma Irradiated |
| Capacity (Metric) | 1 L |