Learn More
Manutan Single sided trolley with 10 x TC6 red bins
Product Code. 12843488
Encompass_Preferred
Change view
Click to view available options
Color:
Red
Product Code. Color
12843488 Red
1 options
Product Code. 12843488 Supplier Manutan Supplier No. A339015
Only null left
Add to basket
Add to basket
Mobile louvre panel trolley, take your bins to where they are needed. Units are powder coated blue with silver/grey base. Supplied with 100mm diameter castors (2 fixed/2 swivel). Non-marking rubber tyres enabling manoeuvrability.
  • Load capacity: 125kg
  • Bins supplied: 10 x TC6 red
  • Dimensions (H x W x D): 1200mm x 1000mm x 500mm
Specifications
Color Red