Learn More
Wesbart™ Single Platform Test Tube Racks
Product Code. 11771724
Click to view available options
Holds:
Tubes (14 mm)
Unit Size:
Each
Product Code. 11771724
Brand: Wesbart™ SP1440
Only null left
Add to basket
Add to basket
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.
  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
Specifications
Product Type Tube Rack
Holds Tubes (14 mm)
Material Aluminum
Positions 40
Diameter (Metric) Opening 14 mm