missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Single Platform Test Tube Rack

Product Code. 16458455
Click to view available options
Holds:
Tubes (18 mm)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16458455

Brand: Wesbart™ SP2412

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Wesbart™ Single Platform Test Tube Racks are ideal for storing short, flat-bottomed test tubes in a freezer or on the bench

  • Designed for use in a horizontal cold store or to be free standing on the bench
  • Labels can be fitted to racks for easy identification
  • Different size test tubes racks are available to suit various needs

Specifications

Product Type Single Platform Test Tube Rack
Array 2 x 6
Autoclavable Autoclavable
Color Natural
Holds Tubes (18 mm)
Material Aluminum
Positions 12
For Use With (Equipment) Upright Freezers
Dimensions (L x W x H) 60 x 180 x 35 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.