Learn More
Epredia™ Single Cytofunnel™ with White Filter Cards
Product Code. 12076679 Shop All Epredia Products
Change view
Click to view available options
Quantity:
50 Pk.
500/Cs.
Unit Size:
Pack of 50
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12076679 50 Pk. Pack of 50
13127963 500/Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12076679 Supplier Epredia™ Supplier No. 5991040
Edge
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
  • Disposable sample chambers with white filter cards and caps
  • Pre-attached filter card
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Quantity 50 Pk.
Type Single Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.1 to 0.5 mL