missing translation for 'onlineSavingsMsg'
Learn More

Manutan Single cutting station, cutting width 1250mm, roll weight 100kg

Product Code. 12876059
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12876059 Supplier Manutan Supplier No. A016025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An ergonomic packaging station especially suitable for lightweight film and corrugated board. Safe bi-directional cutting with adjustable take off height and roller bearing spindle. Each system is supplied with two cartridges, one circular blade, one flat blade. Ergonomically designed cutter cartridge easily replaced in a single operation.

  • Single cutting station with one roll
  • Cutting width: 1000mm
  • Roll weight: 100kg
  • Dimensions (H x W x D): 1165mm x 1370mm x 920mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.