Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand: Thermo Scientific™ 335917000
Code : 04
Additional Details : Weight : 0.80000kg
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Specifications
Adjustable filter holder | |
Measuring transmittance of thin solids, filters and glass or plastic test coupons. Supports filters as small as 10 mm | |
Filters up to 10 mm thick |
Individual Adjustable Filter Holder | |
GENESYS 30/40/50 Spectrophotometers | |
Baseplate for accessory mounting |