Learn More
Biochrom Single Cell Holder
Product Code. 11380382
Change view
Click to view available options
Type:
1-position Cell Holder
Unit Size:
Each
Product Code. Type unitSize
11380382 1-position Cell Holder Each
1 options
Product Code. 11380382 Supplier Biochrom Supplier No. 80210607
Only null left
Add to basket
Add to basket
Single Cell Holder is an accessory for the Ultrospec 7500 and 2100 Pro Spectrophotometers with a 10 to 50 mm pathlength.
  • Single cell holder
  • 10 to 50 mm pathlength
  • Accessory for the Ultrospec 7500, 2100 Pro Spectrophotometer
Specifications
Compatibility Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, 6300
Description Single Cell Holder
Type 1-position Cell Holder
Disposable Reusable
For Use With (Equipment) Ultrospec 7500, 2100 Pro Spectrophotometer
Pathlength 10 to 50 mm
Shape Round