Learn More
R&D Systems™ Single Cell Cartridge Kit for Pala Systems (10 Cartridges)
Product Code. 31781572
Change view
Click to view available options
For Use With (Equipment):
Pala Single Cell Sorter and Dispenser
Unit Size:
1 set
Product Code. For Use With (Equipment) Unit Size
31781572 Pala Single Cell Sorter and Dispenser 1 set
1 options
For research use only. Handle biological samples using appropriate biosafety procedures. Cartridges are single-use and must be disposed of according to laboratory biohazard waste protocols.
Product Code. 31781572 Supplier R&D Systems™ Supplier No. NC003
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For research use only. Handle biological samples using appropriate biosafety procedures. Cartridges are single-use and must be disposed of according to laboratory biohazard waste protocols.
Previously ProteinSimple, now a part of R&D Systems. The Single Cell Cartridge Kit for Pala Systems enables sterile, microfluidic isolation and dispensing of single cells with high accuracy and viability. Individually packaged cartridges support reliable cloning and single-cell workflows.
  • Sterile, single-use microfluidic cartridges designed for precise single-cell isolation and dispensing
  • Enables deposition of one viable cell per well into 96- and 384-well plates for clonal expansion
  • Supports high-efficiency monoclonal cell line development and single-cell genomics workflows
  • Microfluidic design ensures gentle handling to preserve viability of sensitive and primary cells
  • Individually packaged cartridges minimize contamination risk and support sterile workflows
  • Optimized for samples with typical concentrations of 5,000 to20,000 cells/mL
  • Processes sample volumes up to approximately 600 μL
  • Provides reliable single-cell dispensing accuracy for downstream analytical confidence
  • Disposable format eliminates cleaning requirements and reduces system downtime
  • Compatible with R&D Systems™ Pala and Hana Single Cell Dispenser systems
  • Supports applications including monoclonal cell line generation, CRISPR workflows, antibody discovery, and single-cell analysis
  • Kit includes 10 cartridges for consistent, repeatable experimental workflows
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cartridge
For Use With (Equipment) Pala Single Cell Sorter and Dispenser
Includes 10 Sterile Single-use Microfluidic Cartridges
Product Line ProteinSimple
Designed for use with Pala systems. Cartridges are sterile and individually packaged for single-use workflows. Performance may vary depending on sample preparation and cell characteristics.