missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Sinefungin
Description
- Sinefungin is a DNA methyltransferase (DNMT) inhibitor (IC50 = 0.1 - 20 μM; Copeland et al.) and an analog of the methyl donor S-adenosyl-L-methionine (SAM; Nolan; Horiuchi et al.)
- Antiparasitic activity against malarial, trypanosomal, and leishmanial species (Nolan; Trager et al.; Dube et al.)
- Inhibits Newcastle disease and vaccinia virus multiplication (Pugh et al.)
- Inhibits pneumococcal biofilm growth in vitro and colonization in an in vivo rat model (Yadav et al.)
Specifications
Specifications
| CAS | 58944-73-3 |
| Synonym | A 9145; Antibiotic 32232RP; Antibiotic A 9145; RP 32232 |
| Quantity | 1 mg |
| Molecular Formula | C15H23N7O5 |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?