missing translation for 'onlineSavingsMsg'
Learn More

Sinefungin, MedChemExpress

Product Code. 16470949
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470949 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16470949 Supplier Medchem Express Supplier No. HY101938
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sinefungin is a potent inhibitor of virion mRNA(guanine-7-)-methyltransferase, mRNA(nucleoside-2'-)-methyltransferase, and viral multiplication. Sinefungin, a SET7/9 inhibitor, ameliorates renal fibrosis by inhibiting H3K4 methylation.

Sinefungin is a potent inhibitor of virion mRNA(guanine-7-)-methyltransferase, mRNA(nucleoside-2'-)-methyltransferase, and viral multiplication. Sinefungin, a SET7/9 inhibitor, ameliorates renal fibrosis by inhibiting H3K4 methylation.

Specifications

Chemical Name or Material Sinefungin
CAS 58944-73-3
Color White
Purity Grade Notes Research
Molecular Formula C15H23N7O5
Quantity 1 mg
Synonym Adenosyl-Ornithine A-9145 Antibiotic 32232RP
Solubility Information H2O : 100 mg/mL (262.20 mM; Need ultrasonic)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1C[C@@H](N)CC[C@H](N)C(O)=O)N2C(N=CN=C3N)=C3N=C2
Molecular Weight (g/mol) 381.39
Formula Weight 381.39
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.