missing translation for 'onlineSavingsMsg'
Learn More

Sinapyl Alcohol (>85%), TRC

Product Code. 30009786
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009786 2.5 mg 2.5mg
30008862 5 mg 5mg
30010290 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009786 Supplier Toronto Research Chemicals Supplier No. TRCS4868232.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-33-7
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
IUPAC Name 4-[(E)-3-hydroxyprop-1-enyl]-2,6-dimethoxyphenol
SMILES COc1cc(\C=C\CO)cc(OC)c1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H14O4/c1-14-9-6-8(4-3-5-12)7-10(15-2)11(9)13/h3-4,6-7,12-13H,5H2,1-2H3/b4-3+
Formula Weight 210.0892
Chemical Name or Material Sinapyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.