missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Sinapinic acid

Product Code. 16499336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499336 Supplier MedChemExpress Supplier No. HYW009732
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sinapinic acid (Sinapic acid) is a phenolic compound isolated from Hydnophytum formicarum Jack.

Rhizome, acts as an inhibitor of HDAC, with an IC50 of 2.27 mM, and also inhibits ACE-I activity. Sinapinic acid posssess potent anti-tumor activity, induces apoptosis of tumor cells. Sinapinic acid shows antioxidant and antidiabetic activities. Sinapinic acid reduces total cholesterol, triglyceride, and HOMA-IR index, and also normalizes some serum parameters of antioxidative abilities and oxidative damage in ovariectomized rats.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 224.21 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 530-59-6
Product Type Natural Products
Purity 98.9%
Solubility DMSO : 100 mg/mL (446.01 mM∣Need ultrasonic) ∣Ethanol : 25 mg/mL (111.50 mM∣Need ultrasonic)
Synonym Sinapic acid
Target HDAC∣Angiotensin-converting Enzyme (ACE)∣Reactive Oxygen Species∣Apoptosis
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.