missing translation for 'onlineSavingsMsg'
Learn More

SIN3B Antibody (2C11), Novus Biologicals™

Product Code. 18390709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390709 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390709 Supplier Novus Biologicals Supplier No. H00023309M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIN3B Monoclonal antibody specifically detects SIN3B in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIN3B
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_056075
Gene Alias Histone deacetylase complex subunit Sin3b, KIAA0700paired amphipathic helix protein Sin3b, SIN3 homolog B, transcription regulator (yeast), SIN3 homolog B, transcriptional regulator, SIN3 homolog B, transcriptional regulator (yeast), Transcriptional corepressor Sin3b
Host Species Mouse
Immunogen SIN3B (NP_056075.1, 1063 a.a. ~ 1160 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HKMVFIVNSEDYMYRRGTLCRAKQVQPLVLLRHHQHFEEWHSRWLEDNVTVEAASLVQDWLMGEEDEDMVPCKTLCETVHVHGLPVTRYRVQYSRRPA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23309
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.