missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin3A-associated protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97744
This item is not returnable.
View return policy
Description
Sin3A-associated protein Polyclonal specifically detects Sin3A-associated protein in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Sin3A-associated protein | |
| Polyclonal | |
| Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_078821.2 | |
| SAP130 | |
| The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Sin3A-associated protein, 130kDa using the numbering given in entry NP_078821.2 (GeneID 79595). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| 130 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP130, mSin3A-associated protein 130, SAP130, SAP130 Sin3A-associated protein, 130kDa, Sin3-associated polypeptide p130 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79595 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction