missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIN3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£400.00
Specifications
| Antigen | SIN3A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SIN3A Polyclonal specifically detects SIN3A in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| SIN3A | |
| Western Blot 0.5 μg/mL, Flow Cytometry, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q96ST3, Q60520 | |
| 25942 | |
| Synthetic peptide corresponding to residues M(1) K R R L D D Q E S D V Y A A Q Q R R(19) of mouse mSin3A. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 145 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A | |
| SIN3A | |
| IgG | |
| Affinity Purified | |
| mSin 3A |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title