missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sin1/MAPKAP1 Polyclonal antibody specifically detects Sin1/MAPKAP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Sin1/MAPKAP1 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: ATVQDMLSSHHYKSFKVSMIHRLRFTTDVQLGISGDKVEIDPVTNQKASTKFWIKQKPISIDSDLLCACDLAEEKSPSHAIFKLTYLSNHDYKHLYFESDAATVNEIVLKVNYILESRASTARADYFAQKQRKLNRRTSFSFQKEKKSGQ |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?