missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin1/MAPKAP1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40424H
This item is not returnable.
View return policy
Description
Sin1/MAPKAP1 Polyclonal antibody specifically detects Sin1/MAPKAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Sin1/MAPKAP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 | |
| The immunogen recognized by this antibody maps to a region between residue 470 and the C-terminus (residue 522) of human stress-activated map kinase interacting protein 1 using the numbering given in entry NP_001006618.1 (GeneID 79109). | |
| 0.1 mL | |
| Autophagy, Cancer, mTOR Pathway, Protein Kinase | |
| 79109 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction