missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody

Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody
Brand: Invitrogen PA572794
This item is not returnable.
View return policy
Description
Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody for ELISA
Specifications
| Sin Nombre Virus Glycoprotein 1 | |
| Polyclonal | |
| Unconjugated | |
| SNVsMgp1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2654026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| SNVsMgp1 | |
| Sin Nombre virus glycoprotein 1 antibody for a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1. | |
| 100 μg | |
| Primary | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction