missing translation for 'onlineSavingsMsg'
Learn More

Simian Immunodeficiency Virus gp120 Antibody [mFluor Violet 610 SE] - (isolate F236), Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-43772MFV610

344.22 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 30281610

  • £417.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Simian Immunodeficiency Virus gp120 Polyclonal antibody specifically detects Simian Immunodeficiency Virus gp120 in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Simian Immunodeficiency Virus gp120
Polyclonal
Western Blot
Rabbit
Antigen and protein A Affinity-purified
RUO
Virus
Purified
Western Blot
mFluor Violet 610 SE
50 mM Sodium Borate
Produced in rabbits immunized with purified, recombinant Simian Immunodeficiency Virus gp120 (isolate F236) (RefSeq#: CAA32487.1; Gln24-Arg531)
0.1 mL
Primary
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.