missing translation for 'onlineSavingsMsg'
Learn More

Simian Immunodeficiency Virus gp120 Antibody - (isolate F236), Novus Biologicals™

Rabbit Polyclonal Antibody

£154.00 - £302.00

Specifications

Antigen Simian Immunodeficiency Virus gp120
Dilution Western Blot 1:2000-1:20000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30281585
View Documents
Novus Biologicals
NBP3-43772-20ul
20 μL
£154.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30281584
View Documents
Novus Biologicals
NBP3-43772-100ul
100 μL
£302.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Simian Immunodeficiency Virus gp120 Polyclonal antibody specifically detects Simian Immunodeficiency Virus gp120 in Virus samples. It is validated for Western Blot
Specifications

Specifications

Simian Immunodeficiency Virus gp120
Western Blot
Unconjugated
Rabbit
Virus
Produced in rabbits immunized with purified, recombinant Simian Immunodeficiency Virus gp120 (isolate F236) (RefSeq#: CAA32487.1; Gln24-Arg531)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:2000-1:20000
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.