missing translation for 'onlineSavingsMsg'
Learn More

Simetryn, TRC

Product Code. 30428361
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428361 250 mg 250mg
30410409 100 mg -
30422495 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428361 Supplier Toronto Research Chemicals Supplier No. TRCS485070250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1014-70-6
Molecular Formula C8 H15 N5 S
Molecular Weight (g/mol) 213.3
Synonym 1,3,5-Triazine-2,4-diamine, N2,N4-diethyl-6-(methylthio)- (ACI), 1,3,5-Triazine-2,4-diamine, N,N'-diethyl-6-(methylthio)- (9CI), N2,N4-Diethyl-6-(methylthio)-1,3,5-triazine-2,4-diamine (ACI), s-Triazine, 2,4-bis(ethylamino)-6-(methylthio)- (6CI, 7CI, 8CI), 2,4-Bis(ethylamino)-6-methylmercapto-s-triazine, 2,4-Bis(ethylamino)-6-methylthio-1,3,5-triazine, 2,4-Diethylamino-6-methylthio-1,3,5-triazine, 2-Methylmercapto-4,6-bis(ethylamino)-s-triazine, 2-Methylthio-4,6-bis(ethylamino)-1,3,5-triazine, 2-Methylthio-4,6-bis(ethylamino)-s-triazine, 2-Methylthio-4-ethylamino-6-ethylamino-s-triazine, 4,6-Bis(ethylamino)-2-methylthio-1,3,5-triazine, 6-Methylthio-2,4-bis(ethylamino)-s-triazine, G 32911, Gybon, N,N'-Diethyl-6-(methylthio)-1,3,5-triazine-2,4-diamine, NSC 163051, Simetryn, Simetryne
IUPAC Name 2-N,4-N-diethyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
SMILES CCNc1nc(NCC)nc(SC)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H15N5S/c1-4-9-6-11-7(10-5-2)13-8(12-6)14-3/h4-5H2,1-3H3,(H2,9,10,11,12,13)
Formula Weight 213.1048
Chemical Name or Material Simetryn
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.