missing translation for 'onlineSavingsMsg'
Learn More

Simetryn, MedChemExpress

Product Code. 16412296
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412296 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412296 Supplier Medchem Express Supplier No. HYB1853
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Simetryn is a herbicide with the LC50 of 16.9-3.70 mg/L (79.3-17.4 microM.

Simetryn is a herbicide with the LC50 of 16.9-3.70 mg/L (79.3-17.4 microM.

Specifications

Chemical Name or Material Simetryn
CAS 1014-70-6
Color White
Purity Grade Notes Research
Molecular Formula C8H15N5S
Quantity 500 mg
Solubility Information DMSO : ≥ 37 mg/mL (173.46 mM)
SMILES CSC1=NC(NCC)=NC(NCC)=N1
Molecular Weight (g/mol) 213.3
Formula Weight 213.3
Percent Purity 99.39%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.