missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIMC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565196
This item is not returnable.
View return policy
Description
Immunogen sequence: YVIDLTRAEGE NRPIATLDLT LEPVTPSQRE PTSLQTCASL SGKAVMEGQV DRSSQPTARR LINSDPVDLD LVEE Highest antigen sequence identity to the following orthologs - mouse 77%, rat 72%.
SIMC1 (SUMO Interacting Motifs Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeSUMO polymer binding. An important paralog of this gene isSKIDA1.
Specifications
| SIMC1 | |
| Polyclonal | |
| Unconjugated | |
| SIMC1 | |
| 4732471D19Rik; C2H5orf25; C4H5orf25; C5orf25; FLJ44216; oocyte maturation associated 1; OOMA1; platform element for inhibition of autolytic degradation; PLEIAD; Simc1; SUMO interacting motifs containing 1; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 375484 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NDZ2 | |
| SIMC1 | |
| Recombinant Human SIMC1. Recombinant protein control fragment (Product #RP-105939). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction