missing translation for 'onlineSavingsMsg'
Learn More

SIMC1 Polyclonal Antibody

Product Code. 15808583
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15808583 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808583 Supplier Invitrogen Supplier No. PA570170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SIMC1 Polyclonal Antibody

SIMC1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SIMC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIMC1
Gene Accession No. E9Q6E9
Gene Alias 4732471D19Rik; C2H5orf25; C4H5orf25; C5orf25; FLJ44216; oocyte maturation associated 1; OOMA1; platform element for inhibition of autolytic degradation; PLEIAD; Simc1; SUMO interacting motifs containing 1; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1
Gene Symbols SIMC1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SIMC1 (aa 1236-1285).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319719
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.