Learn More
Macherey-Nagel™ Silylating Agent BSA
Product Code. 10125881 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Each
Product Code. Quantity unitSize
10125881 10 mL Each
1 options
Product Code. 10125881 Supplier Macherey-Nagel™ Supplier No. 701210.110
Specifications
Item Description Silylating Agent BSA
Quantity 10 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BSA, 1x10 mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N,O-bis-trimethylsilyl-acetamide