missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 2.0mm (0.08in) dia, hard, 99.9% (metals basis)

Product Code. 11364188
Change view
Click to view available options
Quantity:
50 cm
250 cm
1000 cm
Unit Size:
1000cm
250cm
50cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364188 250 cm 250cm
11384188 50 cm 50cm
11374188 1000 cm 1000cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364188 Supplier Thermo Scientific Alfa Aesar Supplier No. 041457.CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 250 cm
Odor Characteristic
Weight ≈32.94g/m
Form Wire
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver wire, 2.0mm (0.08 in.) dia., Hard

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.