missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.1mm (0.004in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048784
Click to view available options
Quantity:
10 m
2 m
50 m
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 2 m
Percent Purity 99.997%
Grade Premion™
Odor Characteristic
Weight ≈0.08g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Silver wire, 0.1mm (0.004 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'