missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.127mm (0.005in) dia, hard, 99.9% (metals basis)

Product Code. 11394178
Change view
Click to view available options
Quantity:
100 m
25 m
Unit Size:
100m
25m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394178 25 m 25m
11384178 100 m 100m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394178 Supplier Thermo Scientific Alfa Aesar Supplier No. 041454.G9
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Especificaciones

Quantity 25 m
Odor Characteristic
Weight ≈0.13g/m
Form Wire
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver wire, 0.127mm (0.005 in.) dia., Hard, Temper: as drawn

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.