missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.05mm (0.002in) dia, annealed, 99.99% (metals basis)

Product Code. 11490780
Change view
Click to view available options
Quantity:
5 m
50 m
2 x 50 m
Unit Size:
5m
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490780 2 x 50 m Pack of 2
11470780 5 m 5m
11480780 50 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490780 Supplier Thermo Scientific Alfa Aesar Supplier No. 044461.J4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

Silver wire finds application in electronics. It is also used in speaker systems for their magnet wire in order to get the best sound available from their speakers. Further, it is used in jewelry making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 2 x 50 m
Odor Characteristic
Weight ≈0.02g/m
Form Wire
Solubility Information Insoluble in water.
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Silver wire, 0.05mm (0.002 in.) dia., Annealed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.