Learn More
Thermo Scientific Alfa Aesar Silver wire, 0.015mm (0.0006 in.) dia., hard, 99.99% (metals basis)
Brand: Thermo Scientific Alfa Aesar 040732.G5
Silver wire, 0.015mm (0.0006 in.) dia., hard, 99.99% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg