missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Description
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Limited shelf life, 60 days after shipment
Specifications
Specifications
| Chemical Name or Material | Silver two part conductive adhesive |
| Name Note | EXP. x X/XX Resin and Hardener |
| Quantity | 25 g |
| MDL Number | MFCD00003397 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction