missing translation for 'onlineSavingsMsg'
Learn More

Silver trifluoromethanesulfonate, 98%

Product Code. 11379339
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379339 10 g 10g
11389339 2 g 2g
11369339 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379339 Supplier Thermo Scientific Alfa Aesar Supplier No. 088722.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 2923-28-6 | CAgF3O3S | 256.93 g/mol

Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2923-28-6
Molecular Formula CAgF3O3S
Molecular Weight (g/mol) 256.93
MDL Number MFCD00013226
InChI Key QRUBYZBWAOOHSV-UHFFFAOYSA-M
Synonym silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
PubChem CID 76223
IUPAC Name silver;trifluoromethanesulfonate
SMILES [Ag+].[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 356°C
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Beilstein EM>10,
Solubility Information Soluble in water and also in most organic solvents.
Formula Weight 256.93
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Silver trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.