missing translation for 'onlineSavingsMsg'
Learn More

Silver thiocyanate, Thermo Scientific Chemicals

Product Code. 11361998
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361998 50 g 50g
11351998 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361998 Supplier Thermo Scientific Alfa Aesar Supplier No. 011897.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Preparation of Nonmetallic Thiocyanates, Analytical Reagent, in pHotographic Emulsions

Silver thiocyanate is applied in the preparation of nonmetallic thiocyanates, analytical reagent, in photographic emulsions and as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1701-93-5
Molecular Formula CHAgNS
Molecular Weight (g/mol) 166.954
MDL Number MFCD00003416
InChI Key KFDDCZCFSUJOBJ-UHFFFAOYSA-N
Synonym silver 1+ ion thiocyanate ion
PubChem CID 16211904
IUPAC Name silver;thiocyanic acid
SMILES C(#N)S.[Ag]

Specifications

Color White
Physical Form Crystalline
Quantity 50 g
Formula Weight 165.95
Odor Odorless
Appearance White crystals
Chemical Name or Material Silver thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.