missing translation for 'onlineSavingsMsg'
Learn More

Silver tetrafluoroborate, 99%

Product Code. 10468223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10468223 1 g Plastic bottle 1g
10296250 10 g Plastic bottle 10g
15740829 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10468223 Supplier Thermo Scientific Acros Supplier No. 197090010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 14104-20-2 | AgBF4 | 194.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14104-20-2
Molecular Formula AgBF4
Molecular Weight (g/mol) 194.67
MDL Number MFCD00003408
InChI Key CCAVYRRHZLAMDJ-UHFFFAOYSA-N
Synonym silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm
PubChem CID 159722
SMILES [Ag+].F[B-](F)(F)F

Specifications

Color Beige
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 43.92% min. (BF4)
Linear Formula AgBF4
Solubility Information Solubility in water: soluble.
IUPAC Name silver;tetrafluoroborate
Formula Weight 194.67
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Silver tetrafluoroborate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.